Subtyping novel zoonotic pathogen Cryptosporidium chipmunk genotype I.
Journal of clinical microbiology (2015-03-13)
Yaqiong Guo, Elizabeth Cebelinski, Christine Matusevich, Kerri A Alderisio, Marianne Lebbad, John McEvoy, Dawn M Roellig, Chunfu Yang, Yaoyu Feng, Lihua Xiao
RESUMEN
Cryptosporidium chipmunk genotype I is an emerging zoonotic pathogen in humans. The lack of subtyping tools makes it impossible to determine the role of zoonotic transmission in epidemiology. To identify potential subtyping markers, we sequenced the genome of a human chipmunk genotype I isolate. Altogether, 9,509,783 bp of assembled sequences in 853 contigs were obtained, with an N50 of 117,886 bp and >200-fold coverage. Based on the whole-genome sequence data, two genetic markers encoding the 60-kDa glycoprotein (gp60) and a mucin protein (ortholog of cgd1_470) were selected for the development of a subtyping tool. The tool was used for characterizing chipmunk genotype I in 25 human specimens from four U.S. states and Sweden, one specimen each from an eastern gray squirrel, a chipmunk, and a deer mouse, and 4 water samples from New York. At the gp60 locus, although different subtypes were seen among the animals, water, and humans, the 15 subtypes identified differed mostly in the numbers of trinucleotide repeats (TCA, TCG, or TCT) in the serine repeat region, with only two single nucleotide polymorphisms in the nonrepeat region. Some geographic differences were found in the subtype distribution of chipmunk genotype I from humans. In contrast, only two subtypes were found at the mucin locus, which differed from each other in the numbers of a 30-bp minisatellite repeat. Thus, Cryptosporidium chipmunk genotype I isolates from humans and wildlife are genetically similar, and zoonotic transmission might play a potential role in human infections.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Cesium chloride, ReagentPlus®, 99.9%
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥98%
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Cesium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99.9%
Sigma-Aldrich
Cesium chloride, ≥99.999% trace metals basis
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Cesium chloride, Grade I, ≥99.0%
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Cesium chloride, optical grade, ≥99.5% trace metals basis
Sigma-Aldrich
Cesium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cesium chloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Cesium chloride, Grade II, ≥98%
Sigma-Aldrich
Cesium chloride, puriss. p.a., ≥99.5%
Sigma-Aldrich
Cesium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis