• Inicio
  • Resultados de la búsqueda
  • Development of a new nanovesicle formulation as transdermal carrier: formulation, physicochemical characterization, permeation studies and anti-inflammatory activity.
Development of a new nanovesicle formulation as transdermal carrier: formulation, physicochemical characterization, permeation studies and anti-inflammatory activity.
Artificial cells, nanomedicine, and biotechnology (2013-08-16)
Praveen Kumar Gaur, Shikha Mishra, Suresh Purohit, Yatendra Kumar, Anil Bhandari
RESUMEN
Ibuprofen is an important NSAID, however, it can cause GI disturbances when given orally, and employment of transdermal route will require permeation enhancer causing skin injury. Drug-loaded nanovesicles of ceramide-2, cholesterol, palmitic acid, and cholesteryl sulfate (ICVG) were formulated and analyzed for physicochemical and permeation properties. Vesicles were formulated using film hydration method and physicochemical parameters, in vitro drug release, and stability were assessed. Further, nanovesicle gels were evaluated against plain gel containing drug (CG) for ex vivo/in vivo drug permeation and anti-inflammatory activity. The developed formulations showed optimal physicochemical profile and ICV-1 gave 97.24% drug release. Drug permeation was between 17.32 and 33.12 μg/cm(2) for ICVG formulations and 0.27 μg/cm(2) for CG. ICVG-1 and CG showed Cmax of 9.6 and 0.7 μg/ml at 8 and 4 h. ICVG-1 showed 19.9 times higher AUC than CG. Edema inhibition was 57.98% during initial hours by ICVG-1. Ratio of ceramide 2 and palmitic acid plays a critical role in drug permeation through stratum corneum. The stability and protective effect of the formulations were due to ceramide content. The composition has an important role in physicochemical properties and drug permeation thereby generating an optimum formulation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
SAFC
Cholesterol, Plant-Derived
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Palmitic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)