Merck
Vitamin D receptor gene polymorphisms in relation to the risk of colorectal cancer in the Polish population.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-09-10)
Izabela Laczmanska, Lukasz Laczmanski, Marek Bebenek, Pawel Karpinski, Halina Czemarmazowicz, David Ramsey, Andrzej Milewicz, Maria M Sasiadek
RESUMEN
The protective effect of vitamin D against several cancers including colorectal cancer is modulated by the vitamin D receptor (VDR) and its ligand, the active form of vitamin D. VDR response has been found to play a role in various genes encoding proteins involved in crucial cellular pathways. Single nucleotide polymorphisms (SNPs) of the VDR gene that modulate its activity are located in the promoter region, exons 2-9, and their vicinity and also in the 3'UTR region. Some of them have been previously studied in relation to cancer susceptibility and prognosis. The aim of our study was to investigate four polymorphisms, BsmI, ApaI, TaqI, and FokI, of the VDR gene in Polish patients with sporadic colorectal cancer and to evaluate their association with susceptibility to cancer. We found a significant association between the BsmI genotype and cancer (individuals with the bb genotype are more susceptible to cancer compared to those with other genotypes, p = 0.025, Fisher's exact test for 2 × 2 table). Also, the TT genotype at TaqI and the AA genotype at ApaI are correlated with a higher risk of cancer (p = 0.00071 and p = 1.0 × 10(-5), respectively). We found relatively strong linkage disequilibrium between the TaqI and ApaI loci (T with A and t with a, respectively). Both of these loci are associated with cancer. We do not observe any such association for the FokI polymorphism. In conclusion, a small modification in VDR expression may play a role in such a multipathway process as tumorigenesis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for TOC analysis
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
Water, HPLC Plus