Merck
Chondrogenesis of mesenchymal stem cells in an osteochondral environment is mediated by the subchondral bone.
Tissue engineering. Part A (2013-08-29)
Marloes L de Vries-van Melle, Roberto Narcisi, Nicole Kops, Wendy J L M Koevoet, P Koen Bos, J Mary Murphy, Jan A N Verhaar, Peter M van der Kraan, Gerjo J V M van Osch
RESUMEN
In articular cartilage repair, cells that will be responsible for the formation of repair tissue are often exposed to an osteochondral environment. To study cartilage repair mechanisms in vitro, we have recently developed a bovine osteochondral biopsy culture model in which cartilage defects can be simulated reproducibly. Using this model, we now aimed at studying the chondrogenic potential of human bone marrow-derived mesenchymal stem cells (hBMSCs) in an osteochondral environment. In contrast to standard in vitro chondrogenesis, it was found that supplementing transforming growth factor beta (TGFβ) to culture medium was not required to induce chondrogenesis of hBMSCs in an osteochondral environment. hBMSC culture in defects created in osteochondral biopsies or in bone-only biopsies resulted in comparable levels of cartilage-related gene expression, whereas culture in cartilage-only biopsies did not induce chondrogenesis. Subcutaneous implantation in nude mice of osteochondral biopsies containing hBMSCs in osteochondral defects resulted in the formation of more cartilaginous tissue than hBMSCs in chondral defects. The subchondral bone secreted TGFβ; however, the observed results could not be attributed to TGFβ, as either capturing TGFβ with an antibody or blocking the canonical TGFβ signaling pathway did not result in significant changes in cartilage-related gene expression of hBMSCs in the osteochondral culture model. Inhibition of BMP signaling did not prevent chondrogenesis. In conclusion, we demonstrate that chondrogenesis of hBMSCs is induced by factors secreted from the bone. We have strong indications that this is not solely mediated by members of the TGFβ family but other, yet unknown, factors originating from the subchondral bone appeared to play a key role.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
L-Proline, certified reference material, TraceCERT®
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)