Merck
  • Inicio
  • Resultados de la búsqueda
  • Persistent organochlorines in 13 shark species from offshore and coastal waters of Korea: Species-specific accumulation and contributing factors.
Persistent organochlorines in 13 shark species from offshore and coastal waters of Korea: Species-specific accumulation and contributing factors.
Ecotoxicology and environmental safety (2015-02-24)
Hyun-Kyung Lee, Yunsun Jeong, Sunggyu Lee, Woochang Jeong, Eun-Jung Choy, Chang-Keun Kang, Won-Chan Lee, Sang-Jo Kim, Hyo-Bang Moon
RESUMEN
Data on persistent organochlorines (OCs) in sharks are scarce. Concentrations of OCs such as polychlorinated biphenyls (PCBs) and organochlorine pesticides (OCPs) were determined in the muscle tissue of 13 shark species (n=105) collected from offshore (Indian and Pacific Oceans) and coastal waters of Korea, to investigate species-specific accumulation of OCs and to assess the potential health risks associated with consumption of shark meat. Overall OC concentrations were highly variable not only among species but also within the same species of shark. The concentrations of PCBs, DDTs, chlordanes, hexachlorobenzene, and heptachlor in all shark species ranged from <LOQ (limit of quantification) to 184 (mean: 35.0), <LOQ to 1135 (58.2), <LOQ to 56.2 (4.31), <LOQ to 18.8 (1.64) and <LOQ to 77.5 (1.37)ng/g lipid weight, respectively. The determined concentrations of PCBs and DDTs in shark in our study were relatively lower than those reported in other studies. Aggressive shark species and species inhabiting the Indian Ocean had the highest levels of OCs. Inter-species differences in the concentrations and accumulation profiles of OCs among shark species could be explained by differences in feeding habit and sampling locations. Several confounding factors such as growth velocity, trophic position, and regional contamination status may affect the bioaccumulation of OCs in sharks. Hazard ratios of non-cancer risk for all the OCs were below one, whereas the hazard ratios of lifetime cancer risks of PCBs and DDTs exceeded one, implying potential carcinogenic effects in the general population in Korea. This is the first report to document the occurrence of OCs in sharks from Korea.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Nonane, ReagentPlus®, 99%
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Supelco
Nonane, analytical standard
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Supelco
Nonane, analytical standard
Sigma-Aldrich
Nonane, anhydrous, ≥99%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
Supelco
Dichloromethane, analytical standard, stabilized
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, anhydrous, 95%
Supelco
Hexane, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer