Merck
Neuropeptide Y family-degrading metallopeptidases in the Tityus serrulatus venom partially blocked by commercial antivenoms.
Toxicological sciences : an official journal of the Society of Toxicology (2014-09-23)
Daniela Cajado Carvalho, Alexandre K Kuniyoshi, Roberto T Kodama, Ana K Oliveira, Solange M T Serrano, Denise V Tambourgi, Fernanda V Portaro
RESUMEN
Accidents caused by scorpions represent a relevant public health issue in Brazil, being more recurring than incidents with snakes and spiders. The main species responsible for this situation is the yellow scorpion, Tityus serrulatus, due especially to the great frequency with which accidents occur and the potential of its venom to induce severe clinical manifestations, even death, mainly among children. Although neurotoxins are well characterized, little information is known about other components of scorpion venoms, such as peptidases, and their effect on envenomation. Previous results from our group showed that the metallopeptidases present in this venom are capable of hydrolyzing the neuropeptide dynorphin 1-13 in vitro, releasing Leu-enkephalin, which may interact with ion channels and promote indirect neurotoxicity. Thus, this study aims to get more information about the effect of toxic peptidase activity present in the venom on biologically active peptides, and to evaluate the in vitro neutralizing potential of commercial antivenoms produced by the Butantan Institute. A set of human bioactive peptides were studied as substrates for the peptidases, and the members of the neuropeptide Y family were found to be the most susceptible ones. All new substrate hydrolyses were totally inhibited by ethylenediaminetetracetic and not blocked by phenylmethanesulfonylfluoride, indicating that metallopeptidases were responsible for the peptidase activity. Also, peptidase activities were only partially inhibited by therapeutic Brazilian scorpion antivenom (SAV) and arachnid antivenom (AAV). The dose-response inhibition by both antivenoms indicates that AAV neutralizes better than SAV at the used doses. These characterizations, unpublished until now, can contribute to the improvement of our knowledge about the venom and envenomation processes by T. serrulatus.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG