Quantitative method for the determination of iso-fludelone (KOS-1803) in human plasma by LC-MS/MS.
Journal of pharmaceutical and biomedical analysis (2014-08-30)
Susan M Christner, Robert A Parise, Erica D Levine, Naiyer A Rizvi, Mrinal M Gounder, Jan H Beumer
RESUMEN
Epothilones are relatively new tubulin-poison anticancer drugs. Iso-fludelone (KOS-1803) is a synthetic third generation epothilone drug discovered at Memorial Sloan Kettering Cancer Center, and currently in phase I clinical trials. We report an LC-MS/MS assay for the sensitive, accurate and precise quantitation of iso-fludelone in 0.2mL of human plasma. Validation was performed according to FDA guidance. The assay comprised of KOS-1724 as the internal standard and an MTBE liquid-liquid extraction with a water wash step. Separation was achieved with an YMC-Pack ODS-AQ column and an isocratic mobile phase of 0.1% formic acid in acetonitrile and water (70:30, v/v) at 0.3mL/min for 4min. Chromatographic separation was followed by electrospray, positive-mode ionization tandem mass spectrometric detection in the multiple reaction monitoring (MRM) mode. The assay was linear from 0.1 to 300ng/mL and was accurate (-9.41 to -7.07%) and precise (1.03-13.7%) which fulfilled FDA criteria for validation. Recovery from plasma was 73.9-79.7% and ion suppression was negligible (-22.8 to -31.3%). Plasma freeze-thaw stability (99.97-105.7%), stability for 11 months at -80°C (94.93-107.9%), and stability for 6h at room temperature (94.75-105.5%) were all acceptable. This assay is currently being applied to quantitate iso-fludelone in clinical samples.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Supelco
Acetonitrile, analytical standard
Supelco
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%