Merck
  • Inicio
  • Resultados de la búsqueda
  • Bone distribution study of anti leprotic drug clofazimine in rat bone marrow cells by a sensitive reverse phase liquid chromatography method.
Bone distribution study of anti leprotic drug clofazimine in rat bone marrow cells by a sensitive reverse phase liquid chromatography method.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-05-06)
Cheruvu Hanumanth Srikanth, Pankaj Joshi, Anil Kumar Bikkasani, Konica Porwal, Jiaur R Gayen
RESUMEN
The aim of the present study was to investigate the distribution of clofazimine (CLF) in rat bone marrow cells by a validated reverse phase high performance liquid chromatography. CLF and chlorzoxazone (I.S) were extracted by liquid-liquid extraction from plasma and rat bone marrow cells. The chromatographic separation was performed in isocratic mode by the mobile phase consisting of 10mM ammonium formate (pH 3.0 with formic acid) and acetonitrile in a ratio of 50:50 (v/v). The method was accurate and precise in the linear range of 15.6-2000.0 ng/mL with a correlation coefficient (r(2)) of 0.996 and 0.995 in rat plasma and bone marrow cells, respectively. After single oral dose of 20mg/kg, the maximum concentration of CLF in plasma and bone marrow cells were obtained at 12h with the concentrations of 593.2 and 915.4 ng/mL, respectively. The AUC0-t and mean elimination half life (t1/2) of CLF in bone marrow cells were 54339.02 ng h/mL and 52.46 h, respectively, which signified the low body clearance and high distribution of CLF in bone marrow cells. The single oral dose pharmacokinetic investigation was confirmed the CLF endure for a long period in rat due to high distribution in various tissues. The developed method was successfully applied to the estimation of the pharmacokinetic parameters of CLF in plasma and bone marrow cells after administration of single oral dose of 20mg/kg to rats.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Methanol, analytical standard
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Clofazimine
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture