Merck
Occurrences of major polybrominated diphenyl ethers (PBDEs) in maternal and fetal cord blood sera in Korea.
The Science of the total environment (2014-03-19)
Gyuyeon Choi, Sungjoo Kim, Sunmi Kim, Sungkyoon Kim, Youngeun Choi, Hai-Joong Kim, Jeong Jae Lee, Su Young Kim, Sunggyu Lee, Hyo-Bang Moon, Sooran Choi, Kyungho Choi, Jeongim Park
RESUMEN
Polybrominated diphenyl ethers (PBDEs) are of growing public health concern because of their potential toxicities which range from endocrine disruption to neurodevelopment. However, information on their exposure among sensitive human populations is limited. The objectives of this study were to determine the levels of major PBDEs in blood sera of pregnant women and their matching newborn infants. For this purpose, a total of 198 maternal blood samples and 118 matching umbilical cord blood samples were collected from four regions of South Korea in 2011, and were determined for 19 PBDE congeners. Various demographic, dietary, and behavioral characteristics were asked in a questionnaire survey. Average concentration of total PBDEs in maternal blood serum was 3.34 ± 8.42 ng/g lipid weight (lw) at delivery and 3.14 ± 7.46 ng/g lw at 6 months of pregnancy, respectively. In cord blood serum, an average of 9.37 ± 12.60 ng/g lw was detected. Among the measured PBDE congeners, BDE-47, BDE-99 and BDE-153 were most dominant in both maternal and cord blood sera. Relatively higher levels of BDE-99 were detected in cord blood serum. Strong positive correlations were detected between maternal and cord blood serum samples, indicating the importance of maternal transfer. Health consequences of transplacental exposure to PBDEs among fetuses and newborn infants warrant further investigation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Nonane, anhydrous, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Nonane, analytical standard
Supelco
Methanol, analytical standard
Supelco
Nonane, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Nonane, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Supelco
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Dichloromethane solution, certified reference material, 200 μg/mL in methanol