Merck
  • Inicio
  • Resultados de la búsqueda
  • Effect of individual Fc methionine oxidation on FcRn binding: Met252 oxidation impairs FcRn binding more profoundly than Met428 oxidation.
Effect of individual Fc methionine oxidation on FcRn binding: Met252 oxidation impairs FcRn binding more profoundly than Met428 oxidation.
Journal of pharmaceutical sciences (2014-09-02)
Xuan Gao, Junyan A Ji, Karthik Veeravalli, Y John Wang, Taylor Zhang, William Mcgreevy, Kai Zheng, Robert F Kelley, Michael W Laird, Jun Liu, Mary Cromwell
RESUMEN
The long serum half-lives of mAbs are conferred by pH-dependent binding of IgG-Fc to the neonatal Fc receptor (FcRn). The Fc region of human IgG1 has three conserved methionine residues, Met252, Met358, and Met428. Recent studies showed oxidation of these Met residues impairs FcRn binding and consequently affects pharmacokinetics of therapeutic antibodies. However, the quantitative effect of individual Met oxidation on Fc-FcRn binding has not been addressed. This information is valuable for defining critical quality attributes. In the present study, two sets of homodimeric site-directed IgG1 mutations were generated to understand how individual Fc Met oxidation affects FcRn binding. The first approach used Met to Leu mutants to block site-specific Met oxidation. In the other approach, Met to Gln mutants were designed to mimic site-specific Met oxidation. Both mutagenesis approaches show that either Met252 or Met428 oxidation alone significantly impairs Fc-FcRn binding. Met252 oxidation has a more deleterious effect on FcRn binding than M428 oxidation, whereas Met428 oxidation has a bigger destabilization effect on the thermal stability. Our results also show that Met358 oxidation does not affect FcRn binding. In addition, our study suggests that Met to Gln mutation may serve as an important tool to understand Met oxidation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Supelco
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis