Merck
Ectomycorrhizal colonization and growth of the hybrid larch F₁ under elevated CO₂ and O₃.
Environmental pollution (Barking, Essex : 1987) (2014-12-19)
Xiaona Wang, Laiye Qu, Qiaozhi Mao, Makoto Watanabe, Yasutomo Hoshika, Akihiro Koyama, Korin Kawaguchi, Yutaka Tamai, Takayoshi Koike
RESUMEN
We studied the colonization of ectomycorrhizal fungi and species abundance of a hybrid larch (F1) under elevated CO₂ and O₃. Two-year-old seedlings were planted in an Open-Top-Chamber system with treatments: Control (O3 < 6 nmol/mol), O₃ (60 nmol/mol), CO₂ (600 μmol/mol), and CO2 + O₃. After two growing seasons, ectomycorrhiza (ECM) colonization and root biomass increased under elevated CO₂. Additionally, O₃ impaired ECM colonization and species richness, and reduced stem biomass. However, there was no clear inhibition of photosynthetic capacity by O₃. Concentrations of Al, Fe, Mo, and P in needles were reduced by O₃, while K and Mg in the roots increased. This might explain the distinct change in ECM colonization rate and diversity. No effects of combined fumigation were observed in any parameters except the P concentration in needles. The tolerance of F1 to O₃ might potentially be related to a shift in ECM community structure.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N