Merck
Peripheral and spinal mechanisms of nociception in a rat reserpine-induced pain model.
Pain (2015-01-20)
Toru Taguchi, Kimiaki Katanosaka, Masaya Yasui, Koei Hayashi, Mai Yamashita, Koji Wakatsuki, Hiroshi Kiyama, Akihiro Yamanaka, Kazue Mizumura
RESUMEN
Chronic widespread pain is a serious medical problem, yet the mechanisms of nociception and pain are poorly understood. Using a reserpine-induced pain model originally reported as a putative animal model for fibromyalgia, this study was undertaken to examine the following: (1) expression of several ion channels responsible for pain, mechanotransduction, and generation/propagation of action potentials in the dorsal root ganglion (DRG), (2) activities of peripheral nociceptive afferents, and (3) alterations in spinal microglial cells. A significant increase in mRNA expression of the acid-sensing ion channel (ASIC)-3 was detected in the DRG, and the behavioral mechanical hyperalgesia was significantly reversed by subcutaneous injection of APETx2, a selective blocker of ASIC3. Single-fiber recordings in vitro revealed facilitated mechanical responses of mechanoresponsive C-fibers both in the skin and muscle although the proportion of mechanoresponsive C-nociceptors was paradoxically decreased. In the spinal dorsal horn, microglial cells labeled with Iba1 immunoreactivity was activated, especially in laminae I-II where the nociceptive input is mainly processed compared with the other laminae. The activated microglia and behavioral hyperalgesia were significantly tranquilized by intraperitoneal injection of minocycline. These results suggest that the increase in ASIC3 in the DRG facilitated mechanical response of the remaining C-nociceptors and that activated spinal microglia may direct to intensify pain in this model. Pain may be further amplified by reserpine-induced dysfunction of the descending pain inhibitory system and by the decrease in peripheral drive to this system resulting from a reduced proportion of mechanoresponsive C-nociceptors.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Reserpine
Sigma-Aldrich
Minocycline hydrochloride, powder
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Triton X-100, laboratory grade
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Reserpine, European Pharmacopoeia (EP) Reference Standard
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Reserpine Standard for LC-MS, analytical standard, for LC-MS