Merck
  • Inicio
  • Resultados de la búsqueda
  • Exploitation of lectinized lipo-polymerosome encapsulated Amphotericin B to target macrophages for effective chemotherapy of visceral leishmaniasis.
Exploitation of lectinized lipo-polymerosome encapsulated Amphotericin B to target macrophages for effective chemotherapy of visceral leishmaniasis.
Bioconjugate chemistry (2014-05-21)
Pramod K Gupta, Shalini Asthana, Anil K Jaiswal, Vivek Kumar, Ashwni K Verma, Prashant Shukla, Pankaj Dwivedi, Anuradha Dube, Prabhat R Mishra
RESUMEN
We have designed lectin functionalized Lipo-polymerosome bearing Amphotericin B (Lec-AmB-L-Psome) for specific internalization via lectin receptors overexpressed on infected macrophages of mononuclear phagocytic system (MPS) for the effective management of intramacrophage diseases such as visceral leishmaniasis. The lipo-polymerosome composed of glycol chitosan-stearic acid copolymer (GC-SA25%) and model lipid cholesterol was surface-functionalized with lectin by the EDC/NHS carbodiimide coupling method. Our designed Lec-AmB-L-Psome showed >2-fold enhanced uptake and significantly higher internalization in macrophages as compared to AmB-L-Psome. Importantly, pharmacokinetic and organ distribution studies illustrate significantly higher accumulation of Lec-AmB-L-Psome in MPS especially in liver, spleen, and lung as compared to AmB-L-Psome, Ambisome, and Fungizone. The IC50 value demonstrated that Lec-AmB-L-Psome has 1.63, 2.23, and 3.43 times higher activity than AmB-L-Psome (p < 0.01), Ambisome (p < 0.05), and Fungizone (p < 0.05), respectively. Additionally, the Lec-AmB-L-Psome showed significantly higher splenic parasite inhibition (78.66 ± 3.08%) compared to Fungizone and Ambisome that caused only 56.54 ± 3.91% (p < 0.05) and 66.46 ± 2.08% (p < 0.05) parasite inhibition, respectively, in Leishmania-infected hamsters. The toxicity profile revealed that Lec-AmB-L-Psome is a safe delivery system with diminished nephrotoxicity which is a limiting factor of Fungizone application. Taken together, these studies suggest that this surface functionalized self-assembled Lec-AmB-L-Psome can introduce a new platform to specifically target macrophages for effective management of intramacrophage diseases.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
N,N-Dimethylacetamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylacetamide-d9, 99 atom % D
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Supelco
N,N-Dimethylacetamide, analytical standard
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for peptide synthesis, ≥99.8% (GC)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
N,N-Dimethylacetamide, puriss. p.a., ≥99.5% (GC)
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylacetamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
SAFC
N,N-Dimethylacetamide
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material