• Inicio
  • Resultados de la búsqueda
  • Cathodic pseudopolarography: a new tool for the identification and quantification of cysteine, cystine and other low molecular weight thiols in seawater.
Cathodic pseudopolarography: a new tool for the identification and quantification of cysteine, cystine and other low molecular weight thiols in seawater.
Analytica chimica acta (2014-07-01)
Luis M Laglera, Javier Downes, Antonio Tovar-Sánchez, Damiano Monticelli
RESUMEN
Thiols are compounds of paramount importance in the cellular metabolism due to their double detoxifying role as radical scavengers and trace metal ligands. However, we have scarce information about their extracellular cycling as limited data are available about their concentration, stability and speciation in the aquatic medium. In natural waters, they form part of the pool of reduced sulfur substance (RSS) whose presence has been documented by voltammetric and chromatographic methods. Traditional use of cathodic stripping voltammetry (CSV) for the analysis of RSS could only give an overall concentration due to the coalescence of their CSV peaks. Recently, it has been shown that the use of multiple deposition potentials could take voltammetry of RSS to a higher level, permitting the identification and quantification of the mixtures of RSS despite showing as a single coalescent peak. Here, due to its similarity with classical pseudopolarography, we propose to rename this analytical strategy as cathodic pseudopolarography (CP) and we present for the first time its use for the analysis of mixes of low molecular weight thiols (LMWT) at the nanomolar level. Despite limitations caused by the identical behavior of some LMWT, the CP allowed to isolate the contribution of cysteine and cystine from a coalescent signal in LMWT mixtures. Sample handling with clean protocols allowed the direct determination of the cystine:cysteine ratio without sample modification. Finally, we show the application of CP to identify LMWT in seawater samples extracted from benthic chambers and suggest future applications in other areas of environmental electroanalysis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
SAFC
L-Cysteine
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
SAFC
L-Cystine
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
SAFC
L-Cystine
Supelco
L-Cysteine, certified reference material, TraceCERT®