Inhibition of Na(+)/K(+)-ATPase and cytotoxicity of a few selected gold(III) complexes.
Journal of inorganic biochemistry (2014-09-01)
Voin Petrović, Sandra Petrović, Gordana Joksić, Jasmina Savić, Mirjana Čolović, Maria Agostina Cinellu, Lara Massai, Luigi Messori, Vesna Vasić
RESUMEN
Na(+)/K(+)-ATPase is in charge of maintaining the ionic and osmotic intracellular balance by using ATP as an energy source to drive excess Na(+) ions out of the cell in exchange for K(+) ions. We explored whether three representative cytotoxic gold(III) compounds might interfere with Na(+)/K(+)-ATPase and cause its inhibition at pharmacologically relevant concentrations. The tested complexes were [Au(bipy)(OH)2][PF6] (bipy=2,2'-bipyridine), [Au(py(dmb)-H)(CH3COO)2] (py(dmb)-H=deprotonated 6-(1,1-dimethylbenzyl)-pyridine), and [Au(bipy(dmb)-H)(OH)][PF6] (bipy(dmb)-H=deprotonated 6-(1,1-dimethylbenzyl)-2,2'-bipyridine). We found that all of them caused a pronounced and similar inhibition of Na(+)/K(+)-ATPase activity. Inhibition was found to be non-competitive and reversible. Remarkably, treatment with cysteine resulted in reversal or prevention of Na(+)/K(+)-ATPase inhibition. It is very likely that the described effects may contribute to the overall cytotoxic profile of these gold complexes.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
L-Cysteine, 97%
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Tin(II) chloride, reagent grade, 98%
Sigma-Aldrich
Ammonium molybdate tetrahydrate, ACS reagent, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Ammonium molybdate, 99.98% trace metals basis
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
SAFC
L-Cysteine
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Ammonium molybdate tetrahydrate, ACS reagent, 99.98% trace metals basis
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Supelco
Acetic acid, analytical standard