A biochemical study of the distribution of collagen and its crosslinks in knee ligaments and the patellar tendon.
Connective tissue research (2014-08-21)
Mitsuru Hanada, Masaaki Takahashi, Daisuke Suzuki, Masashi Abe, Yukihiro Matsuyama
RESUMEN
The purpose of this study was to investigate biochemical differences in collagen crosslinks from different locations within the ligaments and a tendon of the human knee. The anterior cruciate ligament (ACL), posterior cruciate ligament (PCL), medial collateral ligament (MCL), lateral collateral ligament (LCL), and patellar tendon (PT) were obtained from 24 cadavers (13 men and 11 women) whose average age at the time of death was 84.8 years. Ligaments and PT samples were obtained from the femoral and tibial insertions and the midsubstance. Hydroxyproline (Hyp) and collagen crosslinks, including pyridinoline (Pyr) and pentosidine (Pen), were compared among the different sites. The midsubstance Hyp concentration was greater than at the femoral and tibial insertions in the ACL (p = 0.00124 and 0.000255, respectively) and PCL (p = 0.00036 and 0.042, respectively). The Pyr:collagen ratio did not differ among sites in any of the ligaments or PT. The Pen:collagen ratio at the midsubstance was greater than at the femoral and tibial insertions in the ACL (p = 0.00022 and 0.00025, respectively) and LCL (p = 0.000081 and 0.000021, respectively) and was greater at the femoral insertion in the MCL (p = 0.00010). The mature collagen crosslink Pyr was not different in distribution in knee ligaments and the PT. Pen increased at the midsubstance ligaments and the PT. As increased Pen may represent ligament degeneration, this may indicate that degeneration may progress more rapidly at the midsubstance than at the insertion sites of a ligament.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous