Merck
  • Inicio
  • Resultados de la búsqueda
  • MRI-determined total volumes of visceral and subcutaneous abdominal and trunk adipose tissue are differentially and sex-dependently associated with patterns of estimated usual nutrient intake in a northern German population.
MRI-determined total volumes of visceral and subcutaneous abdominal and trunk adipose tissue are differentially and sex-dependently associated with patterns of estimated usual nutrient intake in a northern German population.
The American journal of clinical nutrition (2015-04-04)
Karina Fischer, Daniela Moewes, Manja Koch, Hans-Peter Müller, Gunnar Jacobs, Jan Kassubek, Wolfgang Lieb, Ute Nöthlings
RESUMEN
Visceral (VAT) and subcutaneous abdominal (SAAT) and trunk (STRAT) adipose tissue (AT) have been suggested to be differentially influenced by diet. We investigated whether and to what extent usual patterns of nutrient intake are associated with VAT, SAAT, and STRAT compared with nondietary predictors in northern German adults (n = 583). AT volumes were quantified by magnetic resonance imaging. Nutrient intake was estimated by a 112-item food-frequency questionnaire linked to the German Food Code and Nutrient Database. Exploratory nutrient patterns were derived by principal components analysis (PCA) and partial least-squares regression (PLS) of 87 nutrients. Cross-sectional associations between nutrient patterns, single nutrients, or total energy intake and AT compartments were analyzed by multiple linear regression. Next to sex and age, respectively, which were important nondietary predictors and accounted for more of the variation in VAT (∼13% and ∼4%) than in SAAT or STRAT (both 4-7% and <1%), variation in VAT (16.8% or 17.6%) was explained to a greater extent by 9 or 2 nutrient patterns derived by principal components analysis or partial least-squares regression, respectively, than was variation in SAAT (10.6% or 8.2%) or STRAT (11.5% or 8.6%). Whereas VAT (16.6%) was primarily explained by nutrient quality, SAAT (6.9%) and STRAT (7.4%) were mainly explained by total energy intake. VAT was positively associated with nutrients characteristic of animal (except for dairy) products, including arachidonic acid (standardized β: 0.25; 95% CI: 0.15, 0.34; P < 0.0001), but negatively with dietary fiber, including polypentoses (standardized β: -0.17; 95% CI: -0.24, -0.09; P < 0.0001), and nutrients found in milk. The direction and strength of many associations, however, depended strongly on sex and adjustment for BMI. VAT may be particularly associated with sex-specific interplays of nutrients found in animal products and fiber, whereas SAAT and STRAT are associated with total energy intake.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)