• Inicio
  • Resultados de la búsqueda
  • The dual role of sulfur-containing amino acids in the synthesis of IV-VI semiconductor nanocrystals: a mechanochemical approach.
The dual role of sulfur-containing amino acids in the synthesis of IV-VI semiconductor nanocrystals: a mechanochemical approach.
Faraday discussions (2014-11-20)
Peter Baláž, Matej Baláž, Mária Caplovičová, Anna Zorkovská, Lubomír Caplovič, Miroslav Psotka
RESUMEN
PbS@cystine nanocrystals were synthesized mechanochemically, with lead acetate and L-cystine being used as the lead and sulfur precursors, respectively. The resulting nanocrystals are 22-34 nm in size, well-faceted and octahedral in shape. Characterization by XRD, FT-IR, NMR, FE-SEM, EDS, TEM (HRTEM) and surface area measurement methods showed that the particles are single, defect-free crystals with a high crystallinity. Furthermore, the crystals were prepared using a solvent-free procedure that was performed under ambient temperature and atmospheric pressure.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
SAFC
L-Cystine
SAFC
L-Cystine
Supelco
L-Cystine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Cystine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%