Merck
A new acetophenone derivative from flowers of Helichrysum italicum (Roth) Don ssp. italicum.
Fitoterapia (2014-10-07)
Daniela Rigano, Carmen Formisano, Ester Pagano, Felice Senatore, Sonia Piacente, Milena Masullo, Raffaele Capasso, Angelo A Izzo, Francesca Borrelli
RESUMEN
A new acetophenone derivative named gnaphaliol 9-O-propanoate (1) was isolated from the chloroform fraction of EtOH extract of Helichrysum italicum ssp. italicum flowers along with the five known acetophenones 12-acetoxytremetone (2), 13-(2-methylpropanoyloxy)toxol (3), [2,3-dihydro-2-[1-(hydroxymethyl)ethenyl]-5-benzofuranyl]-ethanone (4), 1-[2-[1-[(acetyloxy)methyl]ethenyl]-2,3-dihydro-3-hydroxy-5-benzofuranyl]-ethanone (5) and gnaphaliol (6). The structures of compounds 1-6 were elucidated by extensive spectroscopic methods including 1D- ((1)H and (13)C) and 2D-NMR (DQF-COSY, HSQC, HMBC, TOCSY and ROESY) experiments as well as ESIMS analysis. The isolated compounds were investigated for their cytotoxicity, anti-inflammatory and antioxidant properties. Biological assays on human colonic epithelial cells showed that compound 2 possessed antioxidant effects reducing reactive oxygen species (ROS) production.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Dexamethasone, British Pharmacopoeia (BP) Assay Standard