Merck
Nitrite Control over Dissimilatory Nitrate/Nitrite Reduction Pathways in Shewanella loihica Strain PV-4.
Applied and environmental microbiology (2015-03-15)
Sukhwan Yoon, Robert A Sanford, Frank E Löffler
RESUMEN
Shewanella loihica strain PV-4 harbors both a functional denitrification (NO3 (-)→N2) and a respiratory ammonification (NO3 (-)→NH4 (+)) pathway. Batch and chemostat experiments revealed that NO2 (-) affects pathway selection and the formation of reduced products. Strain PV-4 cells grown with NO2 (-) as the sole electron acceptor produced exclusively NH4 (+). With NO3 (-) as the electron acceptor, denitrification predominated and N2O accounted for ∼90% of reduced products in the presence of acetylene. Chemostat experiments demonstrated that the NO2 (-):NO3 (-) ratio affected the distribution of reduced products, and respiratory ammonification dominated at high NO2 (-):NO3 (-) ratios, whereas low NO2 (-):NO3 (-) ratios favored denitrification. The NO2 (-):NO3 (-) ratios affected nirK transcript abundance, a measure of denitrification activity, in the chemostat experiments, and cells grown at a NO2 (-):NO3 (-) ratio of 3 had ∼37-fold fewer nirK transcripts per cell than cells grown with NO3 (-) as the sole electron acceptor. In contrast, the transcription of nrfA, implicated in NO2 (-)-to-NH4 (+) reduction, remained statistically unchanged under continuous cultivation conditions at NO2 (-):NO3 (-) ratios below 3. At NO2 (-):NO3 (-) ratios above 3, both nirK and nrfA transcript numbers decreased and the chemostat culture washed out, presumably due to NO2 (-) toxicity. These findings implicate NO2 (-) as a relevant modulator of NO3 (-) fate in S. loihica strain PV-4, and, by extension, suggest that NO2 (-) is a relevant determinant for N retention (i.e., ammonification) versus N loss and greenhouse gas emission (i.e., denitrification).
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Nitrous oxide, 99%
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Nitrous oxide, ≥99.998%
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium nitrate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium nitrate, ReagentPlus®, ≥99.0%
Supelco
Sodium nitrate, analytical standard
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Supelco
Sodium nitrite, analytical standard
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Supelco
Nitrite ion standard solution, 0.01 M NO2-, for ion-selective electrodes