  • Search Results
  • Characterization of the rat oral microbiome and the effects of dietary nitrate.
Characterization of the rat oral microbiome and the effects of dietary nitrate.
Free radical biology & medicine (2014-10-12)
Embriette R Hyde, Berkley Luk, Stanley Cron, Lenka Kusic, Tyler McCue, Tonya Bauch, Heidi Kaplan, Gena Tribble, Joseph F Petrosino, Nathan S Bryan
ABSTRACT
The nitrate-nitrite-NO pathway to nitric oxide (NO) production is a symbiotic pathway in mammals that is dependent on nitrate reducing oral commensal bacteria. Studies suggest that by contributing NO to the mammalian host, the oral microbiome helps maintain cardiovascular health. To begin to understand how changes in oral microbiota affect physiological functions such as blood pressure, we have characterized the Wistar rat nitrate reducing oral microbiome. Using 16S rRNA gene sequencing and analysis we compare the native Wistar rat tongue microbiome to that of healthy humans and to that of rats with sodium nitrate and chlorhexidine mouthwash treatments. We demonstrate that the rat tongue microbiome is less diverse than the human tongue microbiome, but that the physiological activity is comparable, as sodium nitrate supplementation significantly lowered diastolic blood pressure in Wistar rats and also lowers blood pressure (diastolic and systolic) in humans. We also show for the first time that sodium nitrate supplementation alters the abundance of specific bacterial species on the tongue. Our results suggest that the changes in oral nitrate reducing bacteria may affect nitric oxide availability and physiological functions such as blood pressure. Understanding individual changes in human oral microbiome may offer novel dietary approaches to restore NO availability and blood pressure.
Product Description
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium nitrate, ReagentPlus®, ≥99.0%
Supelco
Sodium nitrate, analytical standard
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium nitrate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis