Merck
  • Inicio
  • Resultados de la búsqueda
  • Isosorbide dinitrate inhibits mechanical stress-induced cardiac hypertrophy and autophagy through downregulation of angiotensin II type 1 receptor.
Isosorbide dinitrate inhibits mechanical stress-induced cardiac hypertrophy and autophagy through downregulation of angiotensin II type 1 receptor.
Journal of cardiovascular pharmacology (2014-06-03)
Li Lin, Jianfeng Xu, Yong Ye, Junbo Ge, Yunzeng Zou, Xuebo Liu
RESUMEN
Mechanical stress can induce cardiac hypertrophy and autophagy. Recently, it has been reported that nitric oxide donors inhibited autophagy in human chondrocytes. Therefore, the effect of isosorbide dinitrate (ISDN) on cardiac hypertrophy and autophagy induced by mechanical stress was investigated in this study. A 48-hour mechanical stretch and a 4-week transverse aortic constriction were performed to induce cardiomyocyte hypertrophy in vitro and in vivo, respectively, before the assessment of myocardial autophagy using LC3b-II. ISDN was found to significantly reduce mechanical stretch-induced LC3b-II upregulation. Furthermore, mechanical stress was shown to upregulate angiotensin II (AngII) type 1 (AT1) receptor expression in both cultured cardiomyocytes and in mouse hearts, whereas ISDN was demonstrated to significantly suppress the upregulation of the AT1 receptor. It was concluded that ISDN could inhibit mechanical stress-induced cardiac hypertrophy and autophagy through the downregulation of AT1 receptor expression.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one, powder
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Supelco
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate sodium salt, ≥99% (HPLC), powder