Analysis of Lujo virus cell entry using pseudotype vesicular stomatitis virus.
Journal of virology (2014-04-18)
Hideki Tani, Koichiro Iha, Masayuki Shimojima, Shuetsu Fukushi, Satoshi Taniguchi, Tomoki Yoshikawa, Yoshihiro Kawaoka, Naoe Nakasone, Haruaki Ninomiya, Masayuki Saijo, Shigeru Morikawa
RESUMEN
Several arenaviruses are known to cause viral hemorrhagic fever (VHF) in sub-Saharan Africa and South America, where VHF is a major public health and medical concern. The biosafety level 4 categorization of these arenaviruses restricts their use and has impeded biological studies, including therapeutic drug and/or vaccine development. Due to difficulties associated with handling live viruses, pseudotype viruses, which transiently bear arenavirus envelope proteins based on vesicular stomatitis virus (VSV) or retrovirus, have been developed as surrogate virus systems. Here, we report the development of a pseudotype VSV bearing each envelope protein of various species of arenaviruses (AREpv), including the newly identified Lujo virus (LUJV) and Chapare virus. Pseudotype arenaviruses generated in 293T cells exhibited high infectivity in various mammalian cell lines. The infections by New World and Old World AREpv were dependent on their receptors (human transferrin receptor 1 [hTfR1] and α-dystroglycan [αDG], respectively). However, infection by pseudotype VSV bearing the LUJV envelope protein (LUJpv) occurred independently of hTfR1 and αDG, indicating that LUJpv utilizes an unidentified receptor. The pH-dependent endocytosis of AREpv was confirmed by the use of lysosomotropic agents. The fusion of cells expressing these envelope proteins, except for those expressing the LUJV envelope protein, was induced by transient treatment at low pH values. LUJpv infectivity was inhibited by U18666A, a cholesterol transport inhibitor. Furthermore, the infectivity of LUJpv was significantly decreased in the Niemann-Pick C1 (NPC1)-deficient cell line, suggesting the necessity for NPC1 activity for efficient LUJpv infection. LUJV is a newly identified arenavirus associated with a VHF outbreak in southern Africa. Although cell entry for many arenaviruses has been studied, cell entry for LUJV has not been characterized. In this study, we found that LUJpv utilizes neither αDG nor hTfR1 as a receptor and found unique characteristics of LUJV glycoprotein in membrane fusion and cell entry. Proper exclusion of cholesterol or some kinds of lipids may play important roles in LUJpv cell entry.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)