Fucoxanthin induces apoptosis in human cervical cancer cell line HeLa via PI3K/Akt pathway.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-08-13)
Guoliu Ye, Qin Lu, Weidong Zhao, Danli Du, Lijie Jin, Yusheng Liu
RESUMEN
Cervical cancer (CC) is a malignant neoplasm arising from cells originating in the cervix uteri, among the top causes of death from cancer in women. In a gene expression profiling study of metabolic response to treatment, PI3K/Akt signaling pathway are associated with the development of CC. A common mechanism of Akt activation seen in cancer types is alterations in the upstream regulators of Akt such as phosphatidylinositol 3-kinase (PI3K), which is overexpressed in cervical cancer tissues, and leads to phosphorylation of Akt. Both PI3K and Akt inhibitors exist and may be therapeutically valuable. In the present study, we use MTT assay and western blot for the high-throughput screening to select specific inhibitors of PI3K/Akt signaling pathway, and then obtain fucoxanthin. Fucoxanthin is a water-soluble dietary fiber, taken from the unique slimy component of alginic cells. Various studies have pointed out that fucoxanthin is very effective for the treatment of cancer. Our results have shown that fucoxanthin induced a significant apoptosis of HeLa cells, compared with other candidates. After treatment with fucoxanthin for 24 h, the level of phosphorylation was inhibited in a dose-dependent manner, and the proteins of apoptotic markers were changed in HeLa cells. And fucoxanthin could suppress tumor growth in vivo. In addition, the mitochondrial signal transduction pathway maybe was involved in its mechanism and NF-κB activation was decreased after treatment with fucoxanthin. Therefore, fucoxanthin may be used as anti-cervical cancer drugs in the future.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)