Merck
  • Inicio
  • Resultados de la búsqueda
  • Influence of tumor microenvironment on prognosis in colorectal cancer: Tissue architecture-dependent signature of endosialin (TEM-1) and associated proteins.
Influence of tumor microenvironment on prognosis in colorectal cancer: Tissue architecture-dependent signature of endosialin (TEM-1) and associated proteins.
Oncotarget (2014-07-02)
Daniel J O'Shannessy, Elizabeth B Somers, Lakshmi K Chandrasekaran, Nicholas C Nicolaides, Jennifer Bordeaux, Mark D Gustavson
RESUMEN
Tumor survival is influenced by interactions between tumor cells and the stromal microenvironment. One example is Endosialin (Tumor Endothelial Marker-1 (TEM-1) or CD248), which is expressed primarily by cells of mesenchymal origin and some tumor cells. The expression, as a function of architectural masking, of TEM-1 and its pathway-associated proteins was quantified and examined for association with five-year disease-specific survival on a colorectal cancer (CRC) cohort divided into training (n=330) and validation (n=164) sets. Although stromal expression of TEM-1 had prognostic value, a more significant prognostic signature was obtained through linear combination of five compartment-specific expression scores (TEM-1 Stroma, TEM-1 Tumor Vessel, HIF2α Stromal Vessel, Collagen IV Tumor, and Fibronectin Stroma). This resulted in a single continuous risk score (TAPPS: TEM-1 Associated Pathway Prognostic Signature) which was significantly associated with decreased survival on both the training set [HR=1.76 (95%CI: 1.44-2.15); p<0.001] and validation set [HR=1.38 (95%CI: 1.02-1.88); p=0.04]. Importantly, since prognosis is a critical clinical question in Stage II patients, the TAPPS score also significantly predicted survival in the Stage II patient (n=126) cohort [HR=1.75 (95%CI: 1.22-2.52); p=0.002] suggesting the potential of using the TAPPS score to assess overall risk in CRC patients, and specifically in Stage II patients.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Benzhydrazide, 98%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
DAPI, for nucleic acid staining