Merck
Enantiospecific determination of arotinolol in rat plasma by LC-MS/MS: application to a stereoselective pharmacokinetic study.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
Zheyuan Qian, Yanhai Xu, Leyi Zheng, Jingbo Zhang, Zhanying Hong, Xiaohang Shen
RESUMEN
A highly sensitive and selective liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed and fully validated for quantification of arotinolol enantiomers in rat plasma using haloperidol as the internal standard. After solid phase extraction of 50.0 μL rat plasma in 96 well plate, a baseline resolution of arotinolol enantiomers was achieved on a CHIRALPAK AD-H column using the mobile phase of n-hexane and ethanol in 0.02% diethylamine (20:80, v/v) at a flow rate of 0.550 mL/min within 11.0 min. Acquisition of mass spectrometric data was performed on a triple-quadrupole mass spectrometer in multiple-reaction-monitoring (MRM) mode with an ESI source using the transition m/z 372.1 → 316.1 for (±)-arotinolol and m/z 376.1 → 165.1 for haloperidol. The calibration curves of both enantiomers were linear over the range of 1.00-200.0 ng/mL (r(2)>0.992) and the lower limit of quantification was 1.00 ng/mL. Intra- and inter-day precision ranged from 5.6% to 8.9% for R-(-)-arotinolol and 4.6-7.4% for S-(+)-arotinolol. Accuracy varied from 0.0% to 7.0% for R-(-)-arotinolol and 5.0-10.0% for S-(+)-arotinolol. For R-(-)-arotinolol, the recovery ranged from 87.2% to 99.2% and the matrix factor was 1.03-1.09; for S-(+)-arotinolol, the recovery ranged from 88.0% to 92.4% and the matrix factor was 0.84-0.95, both were not concentration dependent. The method was demonstrated with acceptable accuracy, precision and specificity for the determination of arotinolol enantiomers and has been successfully applied to a stereoselective pharmacokinetic study.
MATERIALES
Referencia del producto
Marca
Descripción del producto
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Ethanol, for residue analysis
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)
Supelco
Hexane, analytical standard
Millipore
Bifido Selective Supplement B, suitable for microbiology