Merck
Identification and validation co-differentially expressed genes with NAFLD and insulin resistance.
Endocrine (2014-04-05)
Xiao-Chen Wang, Xiao-Rong Zhan, Xin-Yu Li, Jun-Jie Yu, Xiao-Min Liu
RESUMEN
Insulin resistance is a major feature and pathogenic factor of nonalcoholic fatty liver disease (NAFLD). Theoretically, genetic variation in candidate genes related to insulin resistance may contribute to the pathogenesis of NAFLD. The purpose of this study was to identify potentially pathogenic genes involved in NAFLD and insulin resistance that have not yet been discovered. This study yielded five important discoveries. 1. A total of 21 co-differentially expressed genes in both the NAFLD and insulin-resistance groups were identified from the pool containing thousands of genes via the significance analysis of microarrays method. 2. MAP kinase-interacting serine/threonine kinase 2 (Mknk2) was the unique gene to be identified that is involved in the insulin signaling pathway and Mitogen Activated Protein Kinase signaling pathway according to the Kyoto Encyclopedia of Genes and Genomes database. 3. Mknk2 mRNA and protein expression were dose-dependently up-regulated by palmitic acid (PA) in mouse primary hepatocytes. 4. Western blotting analysis and quantitative real-time PCR confirmed that Mknk2 affected the expression of acetyl-CoA carboxylases-1 and fatty acid synthase. 5. The inhibition of Mknk2 alleviated PA-induced insulin resistance, whereas the overexpression of Mknk2 resulted in the aggravation of insulin resistance in PA-treated hepatocytes. Therefore, we predict that MKNK2 may be a key protein related to NAFLD and insulin resistance.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Palmitic acid, ≥95%, FCC, FG
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Supelco
Palmitic acid, analytical standard
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Aurintricarboxylic acid, practical grade, ≥85% (titration), powder
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Supelco
Palmitic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Alanine, European Pharmacopoeia (EP) Reference Standard
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
SAFC
L-Glutamine
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aurintricarboxylic acid ammonium salt, powder