Merck
Impact of equine herpesvirus type 1 (EHV-1) infection on the migration of monocytic cells through equine nasal mucosa.
Comparative immunology, microbiology and infectious diseases (2014-12-03)
Hossein Bannazadeh Baghi, Hans J Nauwynck
RESUMEN
The mucosal surfaces are important sites of entry for a majority of pathogens, and viruses in particular. The migration of antigen presenting cells (APCs) from the apical side of the mucosal epithelium to the lymph node is a key event in the development of mucosal immunity during viral infections. However, the mechanism by which viruses utilize the transmigration of these cells to invade the mucosa is largely unexplored. Here, we establish an ex vivo explant model of monocytic cell transmigration across the nasal mucosal epithelium and lamina propria. Equine nasal mucosal CD172a(+) cells (nmCD172a(+) cells), blood-derived monocytes and monocyte-derived DCs (moDCs) were labeled with a fluorescent dye and transferred to the apical part of a polarized mucosal explant. Confocal imaging was used to monitor the migration patterns of monocytic cells and the effect of equine herpesvirus type 1 (EHV-1) on their transmigration. We observed that 16-26% of mock-inoculated nmCD172a(+) cells and moDCs moved into the nasal epithelia, and 1-7% moved further in the lamina propria. The migration of EHV-1 inoculated monocytic cells was not increased in these tissues compared to the mock-inoculated monocytic cells. Immediate early protein positive (IEP(+)) cells were observed beneath the basement membrane (BM) 48 hours post addition (hpa) of moDCs and nmCD172a(+) cells, but not blood-derived monocytes. Together, our finding demonstrate that monocytic cells may become infected with EHV-1 in the respiratory mucosa and transport the virus from the apical side of the epithelium to the lamina propria en route to the lymph and blood circulation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard