• Inicio
  • Resultados de la búsqueda
  • LC-ESI-MS/MS analysis and pharmacokinetics of GP205, an innovative potent macrocyclic inhibitor of hepatitis C virus NS3/4A protease in rats.
LC-ESI-MS/MS analysis and pharmacokinetics of GP205, an innovative potent macrocyclic inhibitor of hepatitis C virus NS3/4A protease in rats.
Molecules (Basel, Switzerland) (2015-03-11)
Nan Yang, Qiushi Sun, Zihua Xu, Xiuyun Wang, Xin Zhao, Yuqing Cao, Li Chen, Guorong Fan
RESUMEN
A high-throughput, sensitive and specific LC-ESI-MS/MS method was established for the quantitative determination of GP205, a potent inhibitor of hepatitis C virus NS3/4A protease, in rat. The analyte was isolated from 25 μL plasma sample by 96-well LLE. Good linearity was achieved within the concentration range of 2-5000 ng/mL (r2 > 0.996). The intra- and inter-day precision was less than 10%. The accuracy ranged from 0.8% to 5.5% for GP205 in quality control samples at three levels. GP205 was stable during the analysis and the storage period. The method was successfully applied to pharmacokinetic studies of GP205 in Sprague-Dawley rats. The pharmacokinetic profiles of GP205 at three dose levels with oral administration and one dose level with intravenous administration were successfully studied for the first time in SD rats, respectively. After single oral administration of GP205 at the doses of 2.5, 5, 10 mg/kg, respectively, Cmax and AUC0-τ were proportional to the doses given. The absolute bioavailability was estimated as 34% based on the AUCs of oral administration at the dose of 5 mg/kg and intravenous administration at the dose of 1 mg/kg. The data presented in this study provides useful information for further study for GP205.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)