Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Lanthanum, ingot (under oil), 99.9% trace rare earth metals basis
Sigma-Aldrich
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Lanthanum, pieces
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max™, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Lanthanum, rod, 50mm, diameter 6.35mm, cast, 99%
Lanthanum, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Lanthanum, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Lanthanum, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Lanthanum, foil, 100x100mm, thickness 0.1mm, as rolled, 99%
Lanthanum, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Lanthanum, foil, 100x100mm, thickness 0.125mm, as rolled, 99%
Lanthanum, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Lanthanum, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
Lanthanum, rod, 100mm, diameter 6.35mm, cast, 99%
Lanthanum, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)