Merck
  • Inicio
  • Resultados de la búsqueda
  • An armed, YB-1-dependent oncolytic adenovirus as a candidate for a combinatorial anti-glioma approach of virotherapy, suicide gene therapy and chemotherapeutic treatment.
An armed, YB-1-dependent oncolytic adenovirus as a candidate for a combinatorial anti-glioma approach of virotherapy, suicide gene therapy and chemotherapeutic treatment.
Cancer gene therapy (2014-12-17)
Y Kostova, K Mantwill, P S Holm, M Anton
RESUMEN
We investigated the novel recombinant oncolytic adenovirus Ad-delo-sr39TK-RGD, armed with a mutant herpes simplex virus type 1 thymidine kinase (HSV1-sr39TK) as a suicide gene, and explored its antitumor efficacy in combination with HSV1-sr39TK/ganciclovir (GCV) gene therapy and temozolomide (TMZ). Ad-delo-sr39TK-RGD is an E1-mutated conditionally replicating adenovirus dependent on the human Y-box binding protein 1 (YB-1). Thus, we utilized the YB-1 dependency of the vector to target human glioma cells in vitro, using two-dimensional cell culture and three-dimensional multicellular spheroids, and demonstrated the strong replication competence and oncolytic potential of the virus. The cytotoxicity mediated by HSV1-sr39TK and its prodrug GCV enhanced the oncolytic effect even at <0.1 μg ml(-1) GCV and induced cell killing of > 95% after adding GCV 0-1 days following infection. An increased bystander effect of viral replication and GCV in co-cultured infected and uninfected cells was observed. Co-administrating Ad-delo-sr39TK-RGD with TMZ and GCV, spheroid growth was reduced drastically. Gamma counting of infected spheroids demonstrated successful accumulation of the radiotracer (18)F-labeled 9-[4-fluoro-3-(hydroxymethyl)butyl]guanine mediated by HSV1-sr39TK. Hence, our results show that the combination of YB-1-dependent virotherapy with suicide genes and TMZ effectively induces glioma cell killing and may allow for in vivo non-invasive imaging within a limited time frame.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications