Photoinduced intercalation and coordination of a dirhodium complex to DNA: dual DNA binding.
ChemMedChem (2014-04-23)
Alycia M Palmer, Scott J Burya, Judith C Gallucci, Claudia Turro
RESUMEN
Two new complexes, cis-H,H-[Rh2 (OCCH3 NH)2 (LL)(CH3 CN)2 ](2+) , where LL=bpy (2, bpy=2,2'-bipyridine) and dppz (3, dppz=dipyrido[3,2-a:2',3'-c]phenazine), were prepared from the reaction of cis-H,H-[Rh2 (OCCH3 NH)2 (CH3 CN)6 ](2+) (1) with the corresponding bidentate ligand. The bpy and dppz ligands chelate to the same rhodium atom and are positioned trans to the amidato N atoms, as determined by the single crystal X-ray structure of 2. Irradiation of 2 and 3 with visible light in water results in the exchange of one CH3 CNeq ligand for an H2 O molecule with quantum yields, Φ400 , of 0.040 and 0.044, respectively (λirr =400 nm). The identities of the photoproducts of 2 and 3 were determined to be cis-H,H-[Rh2 (OCCH3 NH)2 (L)(H2 O)(CH3 CN)](2+) , where L is bpy (4) and dppz (5), respectively. Mobility shift assays show that 4 crosslinks double-stranded DNA, and ESI-MS experiments indicate that both 4 and 5 form covalent adducts with single-stranded DNA. In addition, relative viscosity and 2D NMR experiments show that the dppz ligand of 5 also intercalates into DNA upon irradiation, making 3 a dual-binding agent that both intercalates and covalently binds to DNA upon the absorption of visible light.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
1,10-Phenanthroline-5,6-dione, 97%
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Rhein
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
Supelco
Dichloromethane, analytical standard, stabilized
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Rhein, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Rhein, technical grade
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol