Dietary DHA during development affects depression-like behaviors and biomarkers that emerge after puberty in adolescent rats.
Journal of lipid research (2014-11-21)
Michael J Weiser, Kelly Wynalda, Norman Salem, Christopher M Butt
RESUMEN
DHA is an important omega-3 PUFA that confers neurodevelopmental benefits. Sufficient omega-3 PUFA intake has been associated with improved mood-associated measures in adult humans and rodents, but it is unknown whether DHA specifically influences these benefits. Furthermore, the extent to which development and puberty interact with the maternal diet and the offspring diet to affect mood-related behaviors in adolescence is poorly understood. We sought to address these questions by 1) feeding pregnant rats with diets sufficient or deficient in DHA during gestation and lactation; 2) weaning their male offspring to diets that were sufficient or deficient in DHA; and 3) assessing depression-related behaviors (forced swim test), plasma biomarkers [brain-derived neurotrophic factor (BDNF), serotonin, and melatonin], and brain biomarkers (BDNF) in the offspring before and after puberty. No dietary effects were detected when the offspring were evaluated before puberty. In contrast, after puberty depressive-like behavior and its associated biomarkers were worse in DHA-deficient offspring compared with animals with sufficient levels of DHA. The findings reported here suggest that maintaining sufficient DHA levels throughout development (both pre- and postweaning) may increase resiliency to emotional stressors and decrease susceptibility to mood disorders that commonly arise during adolescence.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
Testosterone, ≥98%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Oleic acid, natural, FCC