• Inicio
  • Resultados de la búsqueda
  • Oxidative protein refolding on size exclusion chromatography at high loading concentrations: fundamental studies and mathematical modeling.
Oxidative protein refolding on size exclusion chromatography at high loading concentrations: fundamental studies and mathematical modeling.
Journal of chromatography. A (2014-12-03)
Pegah Saremirad, Jeffery A Wood, Yan Zhang, Ajay K Ray
RESUMEN
Size exclusion chromatography has been demonstrated as an effective method for refolding a variety of proteins. However, to date process development mainly relies on laboratory experimentation of individual factors. A robust model is essential for high-throughput process screening and optimization of systems to provide higher productivity and refolding yield. In this work, a detailed kinetic scheme of oxidative refolding of a model protein (lysozyme) has been investigated to predict the refolding results in SEC. Non-reactive native, quenched and equilibrium studies were conducted to obtain the model parameters for the species formed during refolding of denatured/reduced lysozyme. The model was tested in various operating conditions, such as: protein loading concentration, injection volume, flow rate and composition of refolding buffer with and without the use of l-arginine additive. An apparent two-state mechanism was found adequate to describe refolding of lysozyme on SEC for the operating condition tested in this work. Furthermore, using low concentration of l-arginine combined with urea as common aggregation suppressor additives showed insignificant change in kinetics of refolding of lysozyme on SEC. However, addition of l-arginine changed mass transfer properties of some of the species formed in refolding reaction which was considered in the model to accurately predict the result of refolding on SEC.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%