Merck
  • Inicio
  • Resultados de la búsqueda
  • Bioaccumulation and oxidative stress responses measured in the estuarine ragworm (Nereis diversicolor) exposed to dissolved, nano- and bulk-sized silver.
Bioaccumulation and oxidative stress responses measured in the estuarine ragworm (Nereis diversicolor) exposed to dissolved, nano- and bulk-sized silver.
Environmental pollution (Barking, Essex : 1987) (2015-01-01)
Margherita Cozzari, Antonia Concetta Elia, Nicole Pacini, Brian D Smith, David Boyle, Philip S Rainbow, Farhan R Khan
RESUMEN
The impact of Ag NPs on sediment-dwelling organisms has received relatively little attention, particularly in linking bioaccumulation to oxidative injury. The polychaete Nereis diversicolor was exposed to sediments spiked with dissolved Ag (added as AgNO3), Ag NPs (63 ± 27 nm) and larger bulk Ag particles (202 ± 56 μm), for up to 11 days at sublethal concentrations (nominally 2.5, 5, 10 μg Ag g(-1) sediment (dw)). There were concentration- and time-dependent differences in the accumulation of the three Ag forms, but all three forms elicited an oxidative stress response. In the cases of Ag NPs and bulk Ag particles, changes in antioxidant markers (glutathione, SOD, CAT, GPx, SeGPx, GST and GR) occurred without significant Ag accumulation. Differences in biomarker profiles between the three Ag forms suggest that the mechanism of oxidative stress caused by particulate Ag is distinct from that of dissolved Ag.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%