• Inicio
  • Resultados de la búsqueda
  • Antitumor activity of triterpenoid saponin-rich Adisia gigantifolia extract on human breast adenocarcinoma cells in vitro and in vivo.
Antitumor activity of triterpenoid saponin-rich Adisia gigantifolia extract on human breast adenocarcinoma cells in vitro and in vivo.
Biological & pharmaceutical bulletin (2014-06-03)
Li-Hua Mu, Lin Bai, Xian-Zhe Dong, Fu-Qin Yan, Dai-Hong Guo, Xiao-Li Zheng, Ping Liu
RESUMEN
The aim of this study was to explore whether the ethanolic extract of Ardisia gigantifolia rhizomes (AGB-5), a traditional herbal medicine from China, could affect the proliferation of human breast adenocarcinoma (MCF-7) cells in vitro and to explore the antitumor effects of AGB-5 in BALB/c mice engrafted with MCF-7 cells. The results showed that AGB-5 markedly inhibited the proliferation of MCF-7 cells with an IC50 value of 11.89±1.12 µg/mL, increased the S phase and decreased the G2/M phase without influence on G1 phase. MCF-7 cells treated with AGB-5 presented a dose-dependent increase of apoptosis compared with the control group. AGB-5 also significantly increased the activity of caspase-3 and -9 in a dose-dependent manner in MCF-7 cells. Furthermore, in an in vivo model, AGB-5 reduced tumor volume, brought back the red blood cell (RBC) and white blood cell (WBC) count near to normal value, enhanced superoxide dismutase and catalase level of MCF-7 bearing mice. This is the first study to verify the antitumor activity of A. gigantifolia in vivo. The results suggest that AGB-5 may have potential beneficial effects against human breast adenocarcinoma.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Methanol, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Hematoxylin
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder