  • Search Results
  • Modulation of imatinib cytotoxicity by selenite in HCT116 colorectal cancer cells.
Modulation of imatinib cytotoxicity by selenite in HCT116 colorectal cancer cells.
Basic & clinical pharmacology & toxicology (2014-06-17)
Amal Kamal Abdel-Aziz, Samar Saad Eldeen Azab, Samar Samir Youssef, Abeer Mostafa El-Sayed, Ebtehal El-Demerdash, Samia Shouman
ABSTRACT
Imatinib is a principal therapeutic agent for targeting colorectal tumours. However, mono-targeting by imatinib does not always achieve complete cancer eradication. Selenite, a well-known chemopreventive agent, is commonly used in cancer patients. In this study, we aimed to explore whether selenite can modulate imatinib cytotoxicity in colorectal cancer cells. HCT116 cells were treated with different concentrations of imatinib and/or selenite for 24, 48 and 72 hr. Imatinib-selenite interaction was analysed using isobologram equation. As indicators of apoptosis, DNA fragmentation, caspase-3 activity, Bcl-2 expression were explored. Autophagic machinery was also checked by visualizing acidic vesicular organelles and measuring Beclin-1 expression. Furthermore, reactive oxygen and nitrogen species were also examined. This study demonstrated that selenite synergistically augmented imatinib cytotoxicity in HCT116 cells as demonstrated by combination and dose reduction indices. Supranutritional dose of selenite when combined with imatinib induced apoptotic machinery by decreasing Bcl-2 expression, increasing caspase-3 activity and subsequently fragmenting DNA and blunted cytoprotective autophagy by decreasing Beclin-1 expression and autophagosomes formation. Moreover, their combination induced cell cycle S-phase block, increased total thiol content and reduced nitric oxide levels. In conclusion, selenite synergizes imatinib cytotoxicity through multi-barrelled molecular targeting, providing a novel therapeutic approach for colorectal cancer.
Sigma-Aldrich
Sodium nitrate, ReagentPlus®, ≥99.0%
Supelco
Sodium nitrate, analytical standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium nitrate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Caspase 3 Assay Kit, Colorimetric
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis