Probiotics prevent Hirschsprung's disease-associated enterocolitis: a prospective multicenter randomized controlled trial.
International journal of colorectal disease (2014-11-06)
Xiaolin Wang, Zhi Li, Zhilin Xu, Zhongrong Wang, Jiexiong Feng
RESUMEN
Enterocolitis (EC) is the most common and serious postoperative complication of Hirschsprung's disease (HD). Probiotics potentially play a protective role in maintaining intestinal mucosal integrity. Based on the beneficial effects of probiotics, we hypothesized that oral probiotics could decrease the incidence and severity of Hirschsprung's disease-associated enterocolitis (HAEC). We conducted a prospective, multicenter, randomized, and controlled trial to assess whether oral probiotics could decrease the incidence and severity of HAEC. HD patients were randomly assigned into the control group and probiotic-treated group. All children in probiotic-treated group were fed with probiotics per day for 4 weeks. In next 3 months, the incidence and severity of HAEC were analyzed. The peripheral blood T lymphocyte subsets and cytokines, including TNF-α, IFN-γ, IL-6, and IL-10, were analyzed by flow cytometry and enzyme immunoassay (EIA). Compared with the control group, the incidence of HAEC in the probiotic-treated group was significantly diminished. The severity of EC was also remarkably decreased. Furthermore, probiotics balanced T lymphocyte subsets. Moreover, pro-inflammatory cytokines TNF-α, IFN-γ, and IL-6 were significantly decreased and anti-inflammatory cytokine IL-10 was notably increased in probiotic-treated group. Probiotics not only significantly diminished the incidence but also decreased the severity of HAEC. Moreover, our study revealed that probiotics decreased pro-inflammatory cytokine and increased anti-inflammatory cytokine and furthermore balanced T lymphocytes (registered with ClinicalTrials.gov, NCT number: NCT01934959).
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis