The chromatin-modifying protein HMGA2 promotes atypical teratoid/rhabdoid cell tumorigenicity.
Journal of neuropathology and experimental neurology (2015-01-13)
Harpreet Kaur, Marianne Hütt-Cabezas, Melanie F Weingart, Jingying Xu, Yasumichi Kuwahara, Anat Erdreich-Epstein, Bernard E Weissman, Charles G Eberhart, Eric H Raabe
RESUMEN
Atypical teratoid/rhabdoid tumor (AT/RT) is an aggressive pediatric central nervous system tumor. The poor prognosis of AT/RT warrants identification of novel therapeutic targets and strategies. High-mobility Group AT-hook 2 (HMGA2) is a developmentally important chromatin-modifying protein that positively regulates tumor growth, self-renewal, and invasion in other cancer types. High-mobility group A2 was recently identified as being upregulated in AT/RT tissue, but the role of HMGA2 in brain tumors remains unknown. We used lentiviral short-hairpin RNA to suppress HMGA2 in AT/RT cell lines and found that loss of HMGA2 led to decreased cell growth, proliferation, and colony formation and increased apoptosis. We also found that suppression of HMGA2 negatively affected in vivo orthotopic xenograft tumor growth, more than doubling median survival of mice from 58 days to 153 days. Our results indicate a role for HMGA2 in AT/RT in vitro and in vivo and demonstrate that HMGA2 is a potential therapeutic target in these lethal pediatric tumors.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
SAFC
HEPES
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C