Merck
Exchange protein directly activated by cAMP 1 promotes autophagy during cardiomyocyte hypertrophy.
Cardiovascular research (2014-11-21)
Anne-Coline Laurent, Malik Bisserier, Alexandre Lucas, Florence Tortosa, Marie Roumieux, Annélie De Régibus, Audrey Swiader, Yannis Sainte-Marie, Christophe Heymes, Cécile Vindis, Frank Lezoualc'h
RESUMEN
Stimulation of β-adrenergic receptors (β-AR) increases cAMP production and contributes to the pathogenesis of cardiac hypertrophy and failure through poorly understood mechanisms. We previously demonstrated that Exchange protein directly activated by cAMP 1 (Epac1)-induced hypertrophy in primary cardiomyocytes. Among the mechanisms triggered by cardiac stress, autophagy has been highlighted as a protective or harmful response. Here, we investigate whether Epac1 promotes cardiac autophagy and how altered autophagy has an impact on Epac1-induced cardiomyocyte hypertrophy. We reported that direct stimulation of Epac1 with the agonist, Sp-8-(4-chlorophenylthio)-2'-O-methyl-cAMP (Sp-8-pCPT) promoted autophagy activation in neonatal cardiomyocytes. Stimulation of β-AR with isoprenaline (ISO) mimicked the effect of Epac1 on autophagy markers. Conversely, the induction of autophagy flux following ISO treatment was prevented in cardiomyocytes pre-treated with a selective inhibitor of Epac1, CE3F4. Importantly, we found that Epac1 deletion in mice protected against β-AR-induced cardiac remodelling and prevented the induction of autophagy. The signalling mechanisms underlying Epac1-induced autophagy involved a Ca(2+)/calmodulin-dependent kinase kinase β (CaMKKβ)/AMP-dependent protein kinase (AMPK) pathway. Finally, we provided evidence that pharmacological inhibition of autophagy using 3-methyladenine (3-MA) or down-regulation of autophagy-related protein 5 (Atg5) significantly potentiated Epac1-promoted cardiomyocyte hypertrophy. Altogether, these findings demonstrate that autophagy is an adaptive response to antagonize Epac1-promoted cardiomyocyte hypertrophy.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
1,2-Bis(2-amino-5-methylphenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, ≥95% (HPLC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O