  • Search Results
  • Differential expression of doublecortin and microglial markers in the rat brain following fractionated irradiation.
Differential expression of doublecortin and microglial markers in the rat brain following fractionated irradiation.
Neurochemical research (2014-12-10)
Sona Balentova, Eva Hajtmanova, Marian Adamkov, Jan Lehotsky
ABSTRACT
Ionizing radiation induces altered brain tissue homeostasis and can lead to morphological and functional deficits. In this study, adult male Wistar rats received whole-body exposure with fractionated doses of gamma rays (a total dose of 5 Gy) and were investigated 30 and 60 days later. Immunohistochemistry and confocal microscopy were used to determine proliferation rate of cells residing or derived from the forebrain anterior subventricular zone (SVZa) and microglia distributed along and/or adjacent to subventricular zone-olfactory bulb axis. Cell counting was performed in four anatomical parts along the well-defined pathway, known as the rostral migratory stream (RMS) represented by the SVZa, vertical arm, elbow and horizontal arm of the RMS. Different spatiotemporal distribution pattern of cell proliferation was seen up to 60 days after irradiation through the migratory pathway. A population of neuroblasts underwent less evident changes up to 60 days after treatment. Fractionated exposure led to decline or loss of resting as well as reactive forms of microglia until 60 days after irradiation. Results showed that altered expression of the SVZa derived cells and ultimative decrease of microglia may contribute to development of radiation-induced late effects.
Product Description
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard