Merck
Successful entrapping of liposomes in glucan particles: an innovative micron-sized carrier to deliver water-soluble molecules.
Molecular pharmaceutics (2014-08-28)
Francesca Garello, Rachele Stefania, Silvio Aime, Enzo Terreno, Daniela Delli Castelli
RESUMEN
Glucan particles (GPs) are monodisperse microspheres derived from baker's yeast and represent an interesting class of microcarriers for theranostic applications as they show a high affinity toward immune system cells. The typical loading strategy was to harness the ability of the molecule to be loaded to interact with nano-/microassembled systems through electrostatic or hydrophobic forces. However, small water-soluble chemicals could not be steadily retained by the leaky shell of GPs. In this work, we propose an alternative loading approach for small water-soluble compounds that is based on their entrapment in the aqueous core of liposomes that are directly formed into the microparticles through the reverse phase evaporation method (REV). The construct obtained may act as biocompatible carrier to deliver and release, even in a triggerable way, bioactive compounds.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphocholine, ≥99%
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N