Partitioning behavior of perfluorinated compounds between sediment and biota in the Pearl River Delta of South China.
Marine pollution bulletin (2014-04-30)
Y G Zhao, H T Wan, M H Wong, Chris K C Wong
RESUMEN
Surface sediment and biota were collected from 12 sampling sites - seven along the Pearl River Delta and five along the Hong Kong coastline. Perfluorinated compound (PFC) concentrations were detected using a high-performance-liquid-chromatogram-tandem-mass-spectrometry system. Analytical results indicated that the total PFC concentrations were in the range of 0.15-3.11 ng/g dry weight in sediments, while the total PFC concentrations in oyster and mussel samples were between 0.46-1.96 and 0.66-3.43 ng/g wet weight, respectively. The major types of PFCs detected in the sediment samples were perfluorooctanesulfonic acid (PFOS) and perfluorobutanoic acid (PFBA), with concentrations ranging from low limits of quantification to 0.86±0.12 ng/g dry weight and 1.50±0.26 ng/g dry weight, respectively. In bivalve samples, PFOS was the dominant contaminant with concentrations ranging from 0.25±0.09 to 0.83±0.12 ng/g wet weight in oysters and 0.41±0.14 to 1.47±0.25 ng/g wet weight in mussels. An increase in PFC concentration was found to be correlated with increased human population density in the study areas.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%