  • Search Results
  • Anti-angiogenic effects of Siegesbeckia glabrescens are mediated by suppression of the Akt and p70S6K-dependent signaling pathways.
Anti-angiogenic effects of Siegesbeckia glabrescens are mediated by suppression of the Akt and p70S6K-dependent signaling pathways.
Oncology reports (2014-12-02)
Hyeon-Ju Kim, Hee-Young Ko, Shin-Wook Choi, Dong-Wan Seo
ABSTRACT
Siegesbeckia glabrescens (SG) Makino (Compositae) has been used as a traditional medicine for the treatment of allergic and inflammatory diseases. In the present study, we report the effects and molecular mechanism of an ethanolic extract of SG on cell proliferation, migration and tube formation in vascular endothelial growth factor-A (VEGF-A)-treated human umbilical vein endothelial cells. SG treatment inhibited VEGF-A-stimulated endothelial cell proliferation through downregulation of cyclin D and upregulation of cyclin-dependent kinase inhibitors such as p27Kip1 and p21WAF1/Cip1. In addition, SG inhibited VEGF‑A-stimulated endothelial cell migration and tube formation. These anti-angiogenic activities of SG were mediated by inactivation of the Akt- and p70S6K-dependent signaling pathways. Collectively, our findings demonstrate the pharmacological roles and molecular mechanism of SG in regulating angiogenic responses and support further evaluation and development of SG as a potential therapeutic agent for the treatment and prevention of angiogenesis-related diseases including cancer.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Rapamycin, VETRANAL®, analytical standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard