Merck
Metformin in vitro and in vivo increases adenosine signaling in rabbit corpora cavernosa.
The journal of sexual medicine (2014-05-23)
Linda Vignozzi, Sandra Filippi, Paolo Comeglio, Ilaria Cellai, Annamaria Morelli, Giulia Rastrelli, Elena Maneschi, Edoardo Mannucci, Mario Maggi
RESUMEN
In subjects with erectile dysfunction responding poorly to sildenafil, metformin was reported to improve erections. The aim of this study is to investigate metformin's mechanism of action on erectile function, particularly focusing on adenosine (ADO) and nitric oxide (NO) signaling in an animal model of high-fat diet (HFD)-induced metabolic syndrome. In vitro contractility studies of penile strips. Penile expression of genes related to ADO or NO signaling was also evaluated. In vitro contractility studies were used to investigate the effect of in vivo and ex vivo metformin administration on ADO- or acetylcholine (Ach)-induced relaxation of penile strips from HFD as compared with animals fed a regular diet (RD). Expression of ADO receptor type 3 (A3 R), ADO deaminase (ADA), AMP deaminase type 1 (AMPD1), and 2 (AMPD2) was decreased in HFD as compared with RD. Accordingly, in HFD the ADO relaxant effect was potentiated as compared with RD (P < 0.02). In vivo metformin treatment in both RD and HFD significantly increased the ADO relaxing effect (P < 0.0001 and P < 0.01, respectively, vs. relative untreated groups) although to a different extent. In fact, the half-maximal inhibitory concentration (IC50 )/IC50 ratio in RD increased fourfold vs. HFD (RD IC50 ratio = 13.75 ± 2.96; HFD IC50 ratio = 2.85 ± 0.52). In corpora cavernosa (CC) from HFD, in vivo metformin (i) normalized A3 R, ADA, and AMPD1; (ii) further decreased AMPD2; (iii) increased dimethylarginine dimethylamino-hydrolase; and (iv) partially restored impaired Ach-induced relaxation. Ex vivo metformin time and dose dependently increased the relaxant effect of ADO in RD. The potentiating effect of metformin on ADO-induced relaxation was significantly reduced by preincubation with NO synthase inhibitor N(ω) -Nitro-L-arginine methyl ester hydrochloride (L-NAME). Interestingly, in vivo testosterone supplementation in HFD rabbits (i) increased penile expression of endothelial NO synthase and AMPD2 and (ii) restored metformin's potentiating effect on ADO-induced relaxation up to RD level. Metformin in vivo and ex vivo increases ADO signaling in CC, most probably interfering with NO formation and ADO breakdown.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet