Merck
Novel in silico-designed estradiol analogues are cytotoxic to a multidrug-resistant cell line at nanomolar concentrations.
Cancer chemotherapy and pharmacology (2014-12-31)
Anne Theron, Renaud Prudent, Elsie Nolte, Iman van den Bout, Rivak Punchoo, Sumari Marais, Peet du Toit, Yvette Hlophe, Dirk van Papendorp, Laurence Lafanechère, Annie Joubert
RESUMEN
2-Methoxyestradiol (2ME) is a promising anti-cancer agent that disrupts the integrity and dynamics of the spindle network. In order to overcome the pharmacokinetic constraints of this compound, a panel of sulphamoylated estradiol analogues were in silico-designed by our laboratory. In this study, we analysed the potential of each analogue to induce cell death on a panel of cancer cell lines. Moreover, the mechanism of action of the most effective compounds was determined. Cytotoxicity screening of the compounds and intermediates was performed on five different cancer cell lines to determine IG50 values. An in vitro tubulin polymerization assay was done to determine the effect of the drugs on tubulin polymerization while their intracellular effects on the microtubule network were assessed by immunofluorescence microscopy. IG50 calculations showed that the sulphamoylated analogues induce cytotoxicity at nanomolar concentrations in all cell lines, including the P-glycoprotein pump overexpressing multidrug-resistant uterine sarcoma cell line. The non-sulphamoylated compounds were only cytotoxic at micromolar ranges, if at all. The sulphamoylated compounds inhibited pure tubulin polymerization in a dose-dependent manner and induced microtubule destruction in cells after 24-h exposure. Results revealed that the novel sulphamoylated 2ME derivatives have potential as anti-cancer drugs, possibly even against chemoresistant cancer cells. These compounds disrupt the intracellular microtubule integrity which leads to mitotic block of the cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycerol, analytical standard
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm