Merck
Peripheral antinociception and anti-inflammatory effects of sulphated polysaccharides from the alga Caulerpa mexicana.
Basic & clinical pharmacology & toxicology (2014-03-29)
José Gerardo Carneiro, José Ariévilo Gurgel Rodrigues, Edfranck de Sousa Oliveira Vanderlei, Ricardo Basto Souza, Ana Luíza Gomes Quinderé, Chistiane Oliveira Coura, Ianna Wivianne Fernandes de Araújo, Hellíada Vasconcelos Chaves, Mirna Marques Bezerra, Norma Maria Barros Benevides
RESUMEN
Sulphated polysaccharides from marine algae are widely used in biotechnological and pharmaceutical areas. In this study, we evaluated the effects of sulphated polysaccharides from the green marine alga Caulerpa mexicana (Cm-SPs) in nociceptive and inflammatory models in rodents. Cm-SPs (10 or 20 mg/kg), administered i.v. in Swiss mice, significantly reduced nociceptive responses, as measured by the number of writhes in response to acetic acid. Cm-SPs (10 or 20 mg/kg) also reduced second-phase responses in the formalin test, but did not exhibit a significant antinociceptive effect in the hot plate test, suggesting that its antinociceptive action occurs through a peripheral mechanism. Cm-SPs (5, 10 or 20 mg/kg), administered s.c. in wistar rats 1 hr before carrageenan, dextran, histamine or serotonin, were tested in paw oedema models. Cm-SPs (10 or 20 mg/kg) reduced carrageenan-induced paw oedema and myeloperoxidase activity in the paw. In addition, Cm-SPs (20 mg/kg) inhibited dextran- or histamine-induced paw oedema, but not serotonin-induced oedema, suggesting that histamine is the major target of Cm-SPs anti-oedematogenic activity. Finally, Cm-SPs (20 mg/kg) administered in mice did not show significant signs of toxicity. In conclusion, Cm-SPs appear to be promising natural modulatory agents for pain and inflammatory conditions.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Cetylpyridinium chloride, meets USP testing specifications
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality