Merck
The large-scale production of an artificial influenza virus-like particle vaccine in silkworm pupae.
Vaccine (2014-12-03)
Kuniaki Nerome, Shigeo Sugita, Kazumichi Kuroda, Toshiharu Hirose, Sayaka Matsuda, Kei Majima, Kazunori Kawasaki, Toshikatsu Shibata, Okti Nadia Poetri, Retno D Soejoedono, Ni L P Ika Mayasari, Srihadi Agungpriyono, Reiko Nerome
RESUMEN
We successfully established a mass production system for an influenza virus-like particle (VLP) vaccine using a synthetic H5 hemagglutinin (HA) gene codon-optimized for the silkworm. A recombinant baculovirus containing the synthetic gene was inoculated into silkworm pupae. Four days after inoculation, the hemagglutination titer in homogenates from infected pupae reached a mean value of 0.8 million hemagglutination units (HAU), approximately 2,000 μg HA protein per pupa, more than 50-fold higher than that produced with an embryonated chicken egg. VLPs ranging from 30 nm to 300 nm in diameter and covered with a large number of spikes were detected in the homogenates. The spikes were approximately 14 nm long, similar to an authentic influenza HA spike. Detailed electron micrographs indicated that the VLP spike density was similar to that of authentic influenza virus particles. The results clearly show that the expression of a single HA gene can efficiently produce VLPs in silkworm pupae. When chickens were immunized with the pupae homogenate, the hemagglutination inhibition titer in their sera reached values of 2,048-8,192 after approximately 1 month. This is the first report demonstrating that a large amount of VLP vaccine could be produced by single synthetic HA gene in silkworm pupae. Our system might be useful for future vaccine development against other viral diseases.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard