Merck
Exercise mitigates diclofenac-induced liver mitochondrial dysfunction.
European journal of clinical investigation (2014-06-04)
Estela Santos-Alves, Ines Marques-Aleixo, Pedro Coxito, Maria M Balça, David Rizo-Roca, Silvia Rocha-Rodrigues, Sandra Martins, Joan R Torrella, Paulo J Oliveira, Antonio J Moreno, José Magalhães, António Ascensão
RESUMEN
Several strategies have been developed to counteract liver injury as a consequence of nonsteroid anti-inflammatory drugs toxicity. Here, we aimed to determine whether physical exercise results in liver mitochondrial protection against in vitro diclofenac toxicity. Male adult Sprague-Dawley rats were divided into sedentary, 12-week endurance training (ET) and voluntary activity (VPA). In vitro liver mitochondrial function as assessed by oxygen consumption, transmembrane electric potential (ΔΨ) and susceptibility to the mitochondrial permeability transition pore (MPTP) was evaluated in the absence and presence of diclofenac. Mitochondrial oxidative stress markers [MnSOD, aconitase, -SH and MDA, SIRT3, p66shc(Ser36)/p66shc ratio] and apoptotic signalling (caspases 3, 8 and 9, Bax, Bcl-2 and CypD) were assessed. Content of OXPHOS components and qualitative liver morphological evaluation were assessed. Despite no effects of ET and VPA on basal liver mitochondrial oxygen consumption or ΔΨ endpoints, exercised animals showed lower susceptibility to MPTP. Diclofenac-induced decrease in ΔΨ, increased state 4 respiration and susceptibility to MPTP opening were all prevented by exercise. Under untreated conditions, VPA group showed higher aconitase activity, while ET decreased MDA and increased Bax content. VPA decreased p66shc(Ser36), complex III and V OXPHOS subunits. Both ET and VPA increased complex IV OXPHOS subunit, and SIRT3 and Bcl-2 content and decreased caspase 9 activity. Unexpectedly, ET and VPA decreased ANT. Both chronic physical exercise models augmented the resistance to in vitro diclofenac-induced mitochondrial alterations, including increased MPTP susceptibility, possibly by modulating oxidative stress and MPTP regulators.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, analytical standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard