Merck
  • Inicio
  • Resultados de la búsqueda
  • Chemical comparison of Tripterygium wilfordii and Tripterygium hypoglaucum based on quantitative analysis and chemometrics methods.
Chemical comparison of Tripterygium wilfordii and Tripterygium hypoglaucum based on quantitative analysis and chemometrics methods.
Journal of pharmaceutical and biomedical analysis (2014-04-04)
Long Guo, Li Duan, Ke Liu, E-Hu Liu, Ping Li
RESUMEN
Tripterygium wilfordii (T. wilfordii) and Tripterygium hypoglaucum (T. hypoglaucum), two commonly used Chinese herbal medicines derived from Tripterygium genus, have been widely used for the treatment of rheumatoid arthritis and other related inflammatory diseases in clinical therapy. In the present study, a rapid resolution liquid chromatography/electrospray ionization tandem mass spectrometry (RRLC-ESI-MS(n)) method has been developed and validated for simultaneous determination of 19 bioactive compounds including four catechins, three sesquiterpene alkaloids, four diterpenoids, and eight triterpenoids in these two similar herbs. The method validation results indicated that the developed method had desirable specificity, linearity, precision and accuracy. Quantitative analysis results showed that there were significant differences in the content of different types of compounds in T. wilfordii and T. hypoglaucum. Moreover, chemometrics methods such as one-way ANOVA, principal component analysis (PCA) and hierarchical clustering analysis (HCA) were performed to compare and discriminate the two Tripterygium herbs based on the quantitative data of analytes, and it was proven straightforward and reliable to differentiate T. wilfordii and T. hypoglaucum samples from different origins. In conclusion, simultaneous quantification of multiple-active component by RRLC-ESI-MS(n) coupled with chemometrics analysis could be a well-acceptable strategy to compare and evaluate the quality of T. wilfordii and T. hypoglaucum.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard